Learn More
Perkin Elmer US LLC Replacement Air Filter for RF Generator Inlet
Supplier:  Perkin Elmer US LLC N0775220
Encompass_Preferred
To filter compressed air for AA and ICP instrumentation. Replaces 00470652 and N0770198.
Catalog No. 50-904-3882