Learn More
Edmund Optics REPL BULB
Catalog No. NC9267959
Encompass
Replacement Bulb, For 150W EKE, Quartz halogen light source color Temperature is 3200 K, 150W EKE bulb lifetime is typically 200 to 10000 hours, Depending on intensity
Catalog No. NC9267959 Supplier Edmund Optics Supplier No. 39606
Replacement Bulb, For 150W EKE, Quartz halogen light source color Temperature is 3200 K, 150W EKE bulb lifetime is typically 200 to 10000 hours, Depending on intensity