Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Replacement RJ-12 Cable for Liqui-Sense Sensor, 6-pin; 3.5 ft
Catalog No. NC1214403
Encompass
Factory calibrated for water. Adjust calibration for other fluids with the sensitivity adjustment on the controller. Polyester pad wipes clean after spills. Hook-and-loop strip holds sensor pad in place. Pad measures 5" L x 2-5/8" W x 1/16" D (12.7 cm L x 6.7 cm W x 0.16 cm D).
Catalog No. NC1214403 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7709502
Factory calibrated for water. Adjust calibration for other fluids with the sensitivity adjustment on the controller. Polyester pad wipes clean after spills. Hook-and-loop strip holds sensor pad in place. Pad measures 5" L x 2-5/8" W x 1/16" D (12.7 cm L x 6.7 cm W x 0.16 cm D).