Learn More
Cole-Parmer Raven Environmental B-10101-19 Replacement centrifuge tubes for 05477-10 and -20
Catalog No. 50632413 Shop All Cole-Parmer Products
Encompass_Preferred
These impact resistant polycarbonate tubes feature a conical, tapered design. For easy measurements, the tubes show the percentage of volume for compacted solids.
Catalog No. 50-632-413 Supplier Cole-Parmer Supplier No. 0547750
These impact resistant polycarbonate tubes feature a conical, tapered design. For easy measurements, the tubes show the percentage of volume for compacted solids.