Learn More
Cole-Parmer Oakton SS Cell Conductivity Probe, Epoxy, K=1.0; 2.5 ft Cable
Catalog No. NC9838718 Shop All Cole-Parmer Products
Encompass_Preferred
Ultem Body, SS Electrodes Conductivity Cell for OAKTON Conductivity and TDS Meters, K=1.0, 2.5' Cable
Catalog No. NC9838718 Supplier Cole-Parmer Supplier No. 3560850
Ultem Body, SS Electrodes Conductivity Cell for OAKTON Conductivity and TDS Meters, K=1.0, 2.5' Cable