Learn More
Cole-Parmer Cole-Parmer Threaded Ultrasonic Probe Tip, Flat; 1/2" Dia
Supplier:  Cole-Parmer 0471040
Encompass_Preferred
The replaceable tip for this threaded probe is fabricated from the titanium alloy, Ti-6Al-4V and autoclavable.
Catalog No. NC0615530