Learn More
Zaiput Flow Technologies REPL MEMBRANES SEP-10 10PK
SDP
Catalog No. NC2253343
Encompass
Pack of 10 replacement membranes for SEP-10 (medium pore size) - Hydrophobic PTFE
This item is not returnable. View return policy
Catalog No. NC2253343 Supplier Zaiput Flow Technologies Supplier No. OB900S10
Pack of 10 replacement membranes for SEP-10 (medium pore size) - Hydrophobic PTFE