Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pressure Lever Cassette/Cartridge, 3-Stop, POM-C
Catalog No. NC9647031
Encompass
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.
Catalog No. NC9647031 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0762464
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.