Learn More
Trajan Scientific REPL NDL SER II NP.5B-7C
SDP
Catalog No. 502530272
Encompass_Preferred
Trajan 0.5uL Plunger-in-Needle Syringes, Diameter Outer Needle: 0.63 mm, 0.02 in., Length Needle: 70 mm, 2.75 in., Needle Gauge: 23 G, Volume: 0.5 uL, Diameter Inner Needle: 0.003 in., 0.1 mm, Diameter Outer: 0.31 in., 8 mm, For Use With: For nanovolume capillary chromatography injection
Catalog No. 50-253-0272 Supplier Trajan Scientific Supplier No. 033057
Trajan 0.5uL Plunger-in-Needle Syringes, Diameter Outer Needle: 0.63 mm, 0.02 in., Length Needle: 70 mm, 2.75 in., Needle Gauge: 23 G, Volume: 0.5 uL, Diameter Inner Needle: 0.003 in., 0.1 mm, Diameter Outer: 0.31 in., 8 mm, For Use With: For nanovolume capillary chromatography injection