Learn More
Trajan Scientific REPL NDL SERII NLL-5/23H 2/PK
SDP
Catalog No. 502530257
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Side Hole, Diameter Inner: 0.32 mm, 0.012 in., Diameter Outer: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Length Needle: 50 mm, 2 in., Needle Point Style: Side Hole or Dome, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock
Catalog No. 50-253-0257 Supplier Trajan Scientific Supplier No. 039803
Add to Cart
Add to Cart
Trajan Replaceable Luer Lock Needles: Side Hole, Diameter Inner: 0.32 mm, 0.012 in., Diameter Outer: 0.63 mm, 0.024 in., Needle Gauge: 23 G, Length Needle: 50 mm, 2 in., Needle Point Style: Side Hole or Dome, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock