Learn More
Trajan Scientific REPL NDL SII 10UL N10-5 5/PK
SDP
Catalog No. 502530265
Encompass_Preferred
SGE Replacement Needles for 10uL Syringes, Diameter Outer: 0.01 in., 0.25 mm, Needle Gauge: 26 G, Length Needle: 2 in., 51 mm, Needle Point Style: Bevel, Diameter Inner: 0.004 in., 0.11 mm, For Use With: For 10uL syringes, Stainless Steel
Catalog No. 50-253-0265 Supplier Trajan Scientific Supplier No. 037110
SGE Replacement Needles for 10uL Syringes, Diameter Outer: 0.01 in., 0.25 mm, Needle Gauge: 26 G, Length Needle: 2 in., 51 mm, Needle Point Style: Bevel, Diameter Inner: 0.004 in., 0.11 mm, For Use With: For 10uL syringes, Stainless Steel