Learn More
Trajan Scientific REPL NDL SII N25/500-WLC 5/PK
SDP
Catalog No. 502530260
Encompass_Preferred
Trajan Replacement Needles for 25-500uL Syringes, Length Needle: 51 mm, 2 in., Diameter Outer: 0.5 mm, 0.019 in., Diameter Inner: 0.2 mm, 0.007 in., Needle Point Style: LC, Needle Gauge: 25 G, For Use With: Chromatography, Stainless Steel, Quantity: 5/Pk.
Catalog No. 50-253-0260 Supplier Trajan Scientific Supplier No. 038260
Trajan Replacement Needles for 25-500uL Syringes, Length Needle: 51 mm, 2 in., Diameter Outer: 0.5 mm, 0.019 in., Diameter Inner: 0.2 mm, 0.007 in., Needle Point Style: LC, Needle Gauge: 25 G, For Use With: Chromatography, Stainless Steel, Quantity: 5/Pk.