Learn More
Trajan Scientific REPL NDL SII NM-1/2.5-5 5/PK
SDP
Catalog No. 502530259
Encompass_Preferred
Trajan Syringe Plunger and Needle Kits, Length Needle: 50 mm, 1.9 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: 1 to 2.5mL syringe, Quantity: 5/Pk., Replacement Syringe Plunger and Needle
Catalog No. 50-253-0259 Supplier Trajan Scientific Supplier No. 039110
Trajan Syringe Plunger and Needle Kits, Length Needle: 50 mm, 1.9 in., Needle Gauge: 23 G, Needle Point Style: Bevel, Diameter Inner: 0.012 in., 0.32 mm, Diameter Outer: 0.024 in., 0.63 mm, For Use With: 1 to 2.5mL syringe, Quantity: 5/Pk., Replacement Syringe Plunger and Needle