Learn More
Trajan Scientific REPL NDL SII NP.5B-OC-10/.23S
SDP
Catalog No. 502530271
Encompass_Preferred
SGE Replacement Needles for 0.5 - 1uL Syringes, For Use With: For 8mm O.D. barrel 0.5uL microvolume syringes, Needle Gauge: 32 G, Length Needle: 4 in., 101.6 mm, Needle Point Style: Sheathed/LC, Diameter Inner: 0.003 in., 0.1 mm, Diameter Outer: 0.009 in., 0.22 mm, Stainless Steel
Catalog No. 50-253-0271 Supplier Trajan Scientific Supplier No. 033610
SGE Replacement Needles for 0.5 - 1uL Syringes, For Use With: For 8mm O.D. barrel 0.5uL microvolume syringes, Needle Gauge: 32 G, Length Needle: 4 in., 101.6 mm, Needle Point Style: Sheathed/LC, Diameter Inner: 0.003 in., 0.1 mm, Diameter Outer: 0.009 in., 0.22 mm, Stainless Steel