Learn More
Gerstel GERSTEL X2 Replacement Needle, Gastight Design, Compatible with TriStar Syringes, 2 Needles/Pack
SDP
Catalog No. NC1971172
Encompass
Introducing the GERSTEL X2 Replacement Needle, designed for precise liquid injections with 25-500 µL GERSTEL TriStar syringes. This gastight needle ensures sample integrity and accuracy, making it essential for demanding scientific applications. Compatible with GERSTEL MPS robotic systems, it stands out with its high-quality, reliable performance. Each pack includes 2 needles, tailored for consistent and effective use in laboratory settings.
Catalog No. NC1971172 Supplier Gerstel Supplier No. 10011150800
Introducing the GERSTEL X2 Replacement Needle, designed for precise liquid injections with 25-500 µL GERSTEL TriStar syringes. This gastight needle ensures sample integrity and accuracy, making it essential for demanding scientific applications. Compatible with GERSTEL MPS robotic systems, it stands out with its high-quality, reliable performance. Each pack includes 2 needles, tailored for consistent and effective use in laboratory settings.