Learn More
Kuhner Shaker Inc REPL SPNG SILICONE SEAL 5/PK
Catalog No. NC2014346
Encompass
5x Replacement spongy silicone seal for sandwich cover for 96 well MTP (CR1296, CR1296b, CR1396a, 1.5 mm holes)
Catalog No. NC2014346 Supplier Kuhner Shaker Inc Supplier No. SMCR0011
5x Replacement spongy silicone seal for sandwich cover for 96 well MTP (CR1296, CR1296b, CR1396a, 1.5 mm holes)