Learn More
Instech Replacement swivel holder for Instech MCLA, SMCLA balance arms.
SDP
Catalog No. 5081056
Encompass
The Instech GIMBAL (often referred to as a replacement swivel holder) is a mounting component designed for use with Instech's MCLA and SMCLA counterbalanced peripheral arms. This holder allows the swivel to tilt and rotate freely in multiple directions, reducing the mechanical stress on the animal's catheter and tether system. By providing this degree of freedom, the gimbal mount helps prevent tangling and ensures smooth fluid delivery during long-term infusion or sampling studies.
Features
  • Application: Swivel mounting for counterbalanced balance arms
  • Compatibility: Specifically for MCLA and SMCLA arm models
  • Function: Multi-axis movement to reduce tether torque and tension
  • Material: Durable construction for stable swivel positioning
  • Design: Allows for quick swivel installation and removal
  • Reliability: Essential for maintaining catheter patency in active animals
Catalog No. 50-810-56 Supplier Instech Supplier No. GIMBAL
 Restricted Item
Add to Cart
Add to Cart
The Instech GIMBAL (often referred to as a replacement swivel holder) is a mounting component designed for use with Instech's MCLA and SMCLA counterbalanced peripheral arms. This holder allows the swivel to tilt and rotate freely in multiple directions, reducing the mechanical stress on the animal's catheter and tether system. By providing this degree of freedom, the gimbal mount helps prevent tangling and ensures smooth fluid delivery during long-term infusion or sampling studies.
Features
  • Application: Swivel mounting for counterbalanced balance arms
  • Compatibility: Specifically for MCLA and SMCLA arm models
  • Function: Multi-axis movement to reduce tether torque and tension
  • Material: Durable construction for stable swivel positioning
  • Design: Allows for quick swivel installation and removal
  • Reliability: Essential for maintaining catheter patency in active animals