Learn More
World Precision Instrument Replaceable Carbon Fiber Tips for 504517
SDP
Catalog No. 502539343
Encompass
Swiss Tweezers with replaceable carbon fiber tips are great for applications requiring non-metalic, non-marring material is required to prevent damage to delicate or sensitive surfaces. The carbon fiber plastic tips offer a combination of precision and gentleness. These are great for handling electronics components that are sensitive to static electricity, because they are non-conductive. The tips on WPI Swiss Tweezers are pointed. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. The handle is manufactured from an anti-magnetic stainless steel allow, which is suitable for autoclaving or chemical sterilization. These surgical forceps are quality Swiss-made instruments. Replacement carbon fiber tips are also available and sold separately.
Catalog No. 50-253-9343 Supplier World Precision Instrument Supplier No. 504527
Swiss Tweezers with replaceable carbon fiber tips are great for applications requiring non-metalic, non-marring material is required to prevent damage to delicate or sensitive surfaces. The carbon fiber plastic tips offer a combination of precision and gentleness. These are great for handling electronics components that are sensitive to static electricity, because they are non-conductive. The tips on WPI Swiss Tweezers are pointed. These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. The handle is manufactured from an anti-magnetic stainless steel allow, which is suitable for autoclaving or chemical sterilization. These surgical forceps are quality Swiss-made instruments. Replacement carbon fiber tips are also available and sold separately.