Learn More
Chemglass Life Sciences Replacement Hose Connection Set
SDP
Catalog No. 50872078
Encompass_Preferred
Replacement Hose Connection Set. Set consists of one each:• PPN tubulation (A)• Silicone rubber seal (B)• Male PPN hose connection (C)• Female PPN connection (D)Replacement silicone seals can be purchased in packages of 10 each.
Catalog No. 50-872-078 Supplier Chemglass Life Sciences Supplier No. CG156301
Replacement Hose Connection Set. Set consists of one each:• PPN tubulation (A)• Silicone rubber seal (B)• Male PPN hose connection (C)• Female PPN connection (D)Replacement silicone seals can be purchased in packages of 10 each.