Learn More
Analytical Sales & Services Replacement 0.5µm PEEK Frits
Catalog No. 5000807047
Encompass
0.5µm PEEK Replacement Frit .062" x .062" x .250"\n\nQty/Pkg: 10
Catalog No. 50-008-07047 Supplier Analytical Sales & Services Supplier No. 49701X
0.5µm PEEK Replacement Frit .062" x .062" x .250"\n\nQty/Pkg: 10