Learn More
Analytical Sales & Services Replacement 2.0µm PEEK Frits
Catalog No. 5000804313
Encompass
2µm Peek Replacement Frit .062" x .062" x .250"\n\nQty/Pkg: 10PK
Catalog No. 50-008-04313 Supplier Analytical Sales & Services Supplier No. 49700X
2µm Peek Replacement Frit .062" x .062" x .250"\n\nQty/Pkg: 10PK