Learn More
Analytical Sales & Services Replacement 2.0µm SS Frits
Catalog No. 5000807041
Encompass
2µm Replacement SS Frits .062" x .062" x .250"\n\nQty/Pkg: 10PK
Catalog No. 50-008-07041 Supplier Analytical Sales & Services Supplier No. 49101X
2µm Replacement SS Frits .062" x .062" x .250"\n\nQty/Pkg: 10PK