Learn More
Cole-Parmer Argos Technologies EVac™ Replacement 4 Liter PP bottle with barbed tube fittings
Catalog No. NC1549102 Shop All Cole-Parmer Products
Encompass_Preferred
Make the most of your aspiration system with extra bottles, replacement parts, and other accessories. This bottle is ideal for use with 13050-31 and includes the cap with stepless barbed fittings.
Catalog No. NC1549102 Supplier Cole-Parmer Supplier No. 1305039
Make the most of your aspiration system with extra bottles, replacement parts, and other accessories. This bottle is ideal for use with 13050-31 and includes the cap with stepless barbed fittings.