Learn More
Perkin Elmer US LLC Replacement Air Intake Filter for AA Ultra Quiet Oil-Less Air Compressor
Supplier:  Perkin Elmer US LLC N0777870
Encompass_Preferred
With the addition of a water separator with a 5 micron particle filter, no liquid transfers into the air supply, providing you with contaminant free air. Each quiet, vibration free compressor comes standard with an internally and externally coated air tank with top drain.
Catalog No. 50-210-4116