Learn More
Analytical Sales & Services Replacement Analytical-03 Resistor Cartridge, Ratio 15:1-250:1
Catalog No. 5000804522
Encompass
Advantage Replacement Analytical-03 Resistor Cartridge for Adjustable Flow Splitters Models 61 & 62 Ratio 15:1-250:1\n\nQty/Pkg: EACH
Catalog No. 50-008-04522 Supplier Analytical Sales & Services Supplier No. 61113
Advantage Replacement Analytical-03 Resistor Cartridge for Adjustable Flow Splitters Models 61 & 62 Ratio 15:1-250:1\n\nQty/Pkg: EACH