Learn More
Andwin Scientific DURACELL Alkaline Batteries, Button type. 1.5V. 110mA hours
SDP
Catalog No. NC9462307
Encompass_Preferred
DURACELL Alkaline Batteries, Button type. 1.5V. 110mA hours. Long-lasting and dependable, even after seven years of storage. Operate reliably in temperatures of -20 to 54degreeC. Economically bulk-packaged.
Catalog No. NC9462307 Supplier Andwin Scientific Supplier No. 13709660
DURACELL Alkaline Batteries, Button type. 1.5V. 110mA hours. Long-lasting and dependable, even after seven years of storage. Operate reliably in temperatures of -20 to 54degreeC. Economically bulk-packaged.