Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Replacement Battery Covers for Label Printers

Catalog No. 19102831 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Gray
Yellow
For Use With (Equipment):
BMP21-LAB Label Printer
BMP21-PLUS Label Printer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
19-102-831 Gray BMP21-LAB Label Printer
19-102-832 Yellow BMP21-PLUS Label Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-102-831 Supplier Brady™ Supplier No. BMP21LABBATTCOV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement battery covers for BMP™21 portable, BMP21-Lab portable and BMP21-Plus portable label printers

  • Plastic cover snaps in place over battery compartment

Specifications

Product Type Replacement Battery Cover
Color Gray
For Use With (Equipment) BMP21-LAB Label Printer
Material Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.