Learn More
Research Products International Corp Autoclavable, Bench Top Biohazard Bags, 8 1/2 x 11 Inch Bags, 0.72 mil Thick, 100 per Package
SDP
Catalog No. 501367705
Encompass
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.
Bag Only
Catalog No. 50-136-7705 Supplier Research Products International Corp Supplier No. 246831
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene. Ensure safe handling and disposal of biohazardous waste. Fits a variety of containers and holders.
Bag Only