Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Replacement Box
GREENER_CHOICESDP

Catalog No. 31502136 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-502-136 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-136 Supplier Synthware™ Supplier No. K130902
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Storage Box, Adjustable Compartments

Specifications

For Use With (Application) Organic Synthesis
For Use With (Equipment) For storage of 14/10 microscale kit components
Material Plastic
Quantity 1 Ea.
Dimensions (L x W x H) 14 x 9 x 2 in.
Product Type Storage Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.