Learn More
Replacement Bulbs for Spectrophotometers
SCI_ED
Catalog No. S42669BLB
Change view
Click to view available options
Light Source:
Halogen
Catalog No. Light Source
S42669BLB Halogen
1 options
Catalog No. S42669BLB Supplier United Products & Instruments, Inc. Supplier No. 1100505
Add to Cart
Add to Cart
Avoid getting sidetracked by a burned-out bulb. For Fisher Scientific™ Educational Spectrophotometer.
Specifications
Light Source Halogen
Voltage 6 V
Wattage 10 W
For Use With (Equipment) Spectrophotometer
Shape G4
Product Type Halogen Bulb