Learn More
Research Products International Corp Replacement Cap for Kimcole Bottle, White, 10/cs
SDP
Catalog No. 501978272
Encompass
Solid top polypropylene GL 45 screw thread caps have internal molded seal rings. Caps can be used with KimCote Media Bottles and Erlenmeyer Flasks. These caps are autoclavable.
Catalog No. 50-197-8272 Supplier Research Products International Corp Supplier No. 218206
Solid top polypropylene GL 45 screw thread caps have internal molded seal rings. Caps can be used with KimCote Media Bottles and Erlenmeyer Flasks. These caps are autoclavable.