Learn More
Structure Probe Inc Replacement High Penetrating Carbide LASER Cutter Wheel (0.05mm-2.0mm) Materials
SDP
Catalog No. NC1866504
Encompass
These high quality replacement cutting wheels are specifically designed to fit the ULTILE line of Precision Glass Cutters. For cutting materials such as glass or silicon, the carbide wheels are an excellent choice.
Catalog No. NC1866504 Supplier Structure Probe Inc Supplier No. 07735AB
These high quality replacement cutting wheels are specifically designed to fit the ULTILE line of Precision Glass Cutters. For cutting materials such as glass or silicon, the carbide wheels are an excellent choice.