Learn More
MSC Extract-All Replacement Carbon Air Filter, Activated Carbon, 15 lbs., 1 ea
Catalog No. 5000508444
Encompass
This replacement carbon filter is designed for use with F-984-5A units. It effectively removes various airborne contaminants to maintain air quality in industrial environments.
  • Features activated carbon media
  • Designed for removal of odors, smoke, and VOCs
  • Weighs 15 lbs.
  • Compatible with specified air filtration equipment
  • Includes 1 filter per package
Catalog No. 50-005-08444 Supplier MSC Supplier No. 60812583
This replacement carbon filter is designed for use with F-984-5A units. It effectively removes various airborne contaminants to maintain air quality in industrial environments.
  • Features activated carbon media
  • Designed for removal of odors, smoke, and VOCs
  • Weighs 15 lbs.
  • Compatible with specified air filtration equipment
  • Includes 1 filter per package