Learn More
Broadley James Corporation REPLACEMENT CARTIDGE 4/PK
Catalog No. NC9928032
Encompass
Kit, Four Cartridge; Contains everything required to replace four membrane cartridges on a 12mm OxyProbe; Includes: One 25mL bottle of electrolyte, Four membrane cartridges, Four sets of internal o-rings
Catalog No. NC9928032 Supplier Broadley James Corporation Supplier No. KA1204
Kit, Four Cartridge; Contains everything required to replace four membrane cartridges on a 12mm OxyProbe; Includes: One 25mL bottle of electrolyte, Four membrane cartridges, Four sets of internal o-rings