Learn More
Optimize Technologies Replacement Cartridge for Active Inlet Check Valve, 600bar HP/Agilent 1050, 1100, 1200
SDP
Catalog No. NC0588228
Encompass_Preferred
The OPTI-MAX 600 bar Active Inlet replacement cartridge (AIV) retails for about the same price as the 400 bar active inlet cartridge offered by Agilent. The OPTI-MAX 600 bar AIV utilizes the proprietary OPTI-MAX technology with wetted materials being 316 SS, PEEK and Zirconia for complete compatibility and reliability in both 400 bar and 600 bar applications. The Zirconia ball/seat sets are individually tested to perform at 3 bubbles or less in the industry-standard 90 PSI nitrogen leak test and withstand well over 20,000 PSI as the assembled cartridge. Ball travel is minimized which allows the OPTI-MAX AIV cartridge to exhibit extremely low pulsation in both 600 bar and 400 bar applications. The rugged 316 SS cartridge body allows the OPTI-MAX AIV cartridge to be easily inserted and tightened for the higher pressure application of 600 bar, while being compatible with both 600 and 400 bar instruments. This item is equivalent to OEM part number G1312-60020 and 5062-8562.
Catalog No. NC0588228 Supplier Optimize Technologies Supplier No. 273804713
The OPTI-MAX 600 bar Active Inlet replacement cartridge (AIV) retails for about the same price as the 400 bar active inlet cartridge offered by Agilent. The OPTI-MAX 600 bar AIV utilizes the proprietary OPTI-MAX technology with wetted materials being 316 SS, PEEK and Zirconia for complete compatibility and reliability in both 400 bar and 600 bar applications. The Zirconia ball/seat sets are individually tested to perform at 3 bubbles or less in the industry-standard 90 PSI nitrogen leak test and withstand well over 20,000 PSI as the assembled cartridge. Ball travel is minimized which allows the OPTI-MAX AIV cartridge to exhibit extremely low pulsation in both 600 bar and 400 bar applications. The rugged 316 SS cartridge body allows the OPTI-MAX AIV cartridge to be easily inserted and tightened for the higher pressure application of 600 bar, while being compatible with both 600 and 400 bar instruments. This item is equivalent to OEM part number G1312-60020 and 5062-8562.