Learn More
Grainger Elga Replacement Cartridge, Mixed Bed Resin, Tap Water, 4/Pk.
Catalog No. 502789852
Encompass_Preferred
This replacement cartridge is constructed from high density polyethylene and contains mixed bed ion exchange resins for water purification applications. It is designed for use with tap water feed sources to effectively remove ions from the water supply.
  • Features mixed bed ion exchange resins for ion removal.
  • Constructed from durable high density polyethylene housing.
  • Designed for tap water purification systems.
  • Easy to replace in compatible water purification equipment.
  • Flow rate of 30 L/hour.
  • Operating temperature range of 0°C to 50°C.
  • Pack contains 4 cartridges.
Catalog No. 50-278-9852 Supplier Grainger Supplier No. 16D239
This replacement cartridge is constructed from high density polyethylene and contains mixed bed ion exchange resins for water purification applications. It is designed for use with tap water feed sources to effectively remove ions from the water supply.
  • Features mixed bed ion exchange resins for ion removal.
  • Constructed from durable high density polyethylene housing.
  • Designed for tap water purification systems.
  • Easy to replace in compatible water purification equipment.
  • Flow rate of 30 L/hour.
  • Operating temperature range of 0°C to 50°C.
  • Pack contains 4 cartridges.