Learn More
Parker Hannifan Parker Hannifin Replacement Catalyst Module, Hydrocarbon Removal, Stainless Steel Vessel, 2 Units
SDP
Catalog No. NC9123881
Encompass
The Parker Hannifin Replacement Catalyst Module significantly enhances air quality by ensuring the thorough removal of hydrocarbons from air supplies in Zero Air Generators. This robust module, designed with a stainless steel vessel and integrated cartridge heater and controller, differentiates itself by effectively transforming harmful hydrocarbons into harmless carbon dioxide and water vapor, thus maintaining compliance with strict removal specifications for uninterrupted generator performance.
Catalog No. NC9123881 Supplier Parker Hannifan Supplier No. 76811
Add to Cart
Add to Cart
Request a Quote
The Parker Hannifin Replacement Catalyst Module significantly enhances air quality by ensuring the thorough removal of hydrocarbons from air supplies in Zero Air Generators. This robust module, designed with a stainless steel vessel and integrated cartridge heater and controller, differentiates itself by effectively transforming harmful hydrocarbons into harmless carbon dioxide and water vapor, thus maintaining compliance with strict removal specifications for uninterrupted generator performance.