Learn More
TA Instruments Replacement Certified Indium Reference Material, Certified Indium, Traceable Temperature Calibration, DSC and TMA Instruments, Documentation from National Metrology Lab
Catalog No. 5000580734 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement Certified Indium Reference Material is a certified indium reference used for traceable temperature calibration of DSC and TMA instruments, supplied with documentation from a national metrology lab.
Catalog No. 50-005-80734 Supplier TA Instruments Supplier No. 915061.901
Waters TA Instruments Replacement Certified Indium Reference Material is a certified indium reference used for traceable temperature calibration of DSC and TMA instruments, supplied with documentation from a national metrology lab.