Learn More
TA Instruments Replacement Certified Nickel Reference Material, Magnetic Reference Material, Temperature Calibration, TGA Instruments, SDT Instruments, Replacement Certified Nickel Reference Material
Catalog No. 5000580615 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement Certified Nickel Reference Material is a magnetic reference material for temperature calibration of TGA and SDT instruments. It serves as a replacement component for maintaining accurate instrument calibration.
Catalog No. 50-005-80615 Supplier TA Instruments Supplier No. 952385.901
Waters TA Instruments Replacement Certified Nickel Reference Material is a magnetic reference material for temperature calibration of TGA and SDT instruments. It serves as a replacement component for maintaining accurate instrument calibration.