Learn More
Perkin Elmer US LLC Replacement Charcoal Trap for Capillary Injector
Catalog No. 502112620
Encompass_Preferred
This trap is packed with activated charcoal and is mainly for removing the trace amounts of hydrocarbons and organics from carrier gases within each sample. Reduces organics to 0.1 ppm (assuming 100 ppm input).
Catalog No. 50-211-2620 Supplier Perkin Elmer US LLC Supplier No. N6100331
This trap is packed with activated charcoal and is mainly for removing the trace amounts of hydrocarbons and organics from carrier gases within each sample. Reduces organics to 0.1 ppm (assuming 100 ppm input).