Learn More
Electron Microscopy Sciences Replacement Chisel
SDP
Catalog No. 50996715
Encompass
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.
Catalog No. 50-996-715 Supplier Electron Microscopy Sciences Supplier No. 6209712
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.