Learn More
Hach Company Replacement conductivity sensor for Sension+ 5049 multisensor
Catalog No. 501997999 Shop All Hach Company Products
Encompass
Laboratory and field probes for use with Sension+ meters.A wide selection of Sension+ electrodes are available to meet almost all laboratory and field applications. Measureable parameters include pH, Dissolved Oxygen, Conductivity, ORP, Ammonia, Ammonium, Chloride, Fluoride, Nitrate, and Sodium. All pH probes are delivered with storage tubes. Field probes are equipped with screwable calibration tubes, serving excellent usability in the field and minimising buffer consumption. Several multi probes are available for Sension+ portable multi meters, saving time and money due to the simultaneous measurement of up to three parameters, combined with convenient handling.
Catalog No. 50-199-7999 Supplier Hach Company Supplier No. LZW9172.99
Laboratory and field probes for use with Sension+ meters.A wide selection of Sension+ electrodes are available to meet almost all laboratory and field applications. Measureable parameters include pH, Dissolved Oxygen, Conductivity, ORP, Ammonia, Ammonium, Chloride, Fluoride, Nitrate, and Sodium. All pH probes are delivered with storage tubes. Field probes are equipped with screwable calibration tubes, serving excellent usability in the field and minimising buffer consumption. Several multi probes are available for Sension+ portable multi meters, saving time and money due to the simultaneous measurement of up to three parameters, combined with convenient handling.