Learn More
Research Products International Corp Replacement Cover for Immuno-Staining Tray, Black
SDP
Catalog No. 501978040
Encompass
Replacement cover for use with Immuno-Staining tray. A black lid for fluorescent work. Made of ABS with a temperature range of -80°C to +80°C
Catalog No. 50-197-8040 Supplier Research Products International Corp Supplier No. 195804
Replacement cover for use with Immuno-Staining tray. A black lid for fluorescent work. Made of ABS with a temperature range of -80°C to +80°C