Learn More
Fisherbrand™ Replacement Deuterium Lamp for Waters UV and Visible HPLC Detector
Catalog No. 14500731 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Waters 2487 HPLC Detector
Catalog No. For Use With (Equipment)
14-500-731 Waters 2487 HPLC Detector
1 options
Catalog No. 14-500-731
Fisherbrand™ Replacement Deuterium Lamp for Waters UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-731 is comparable to Waters part number WAS081142.
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
For Use With (Equipment) Waters 2487 HPLC Detector
Product Type Replacement Detector Lamp