Learn More
Cooper Atkins Corporation REPLACEMENT DURA NEEDLE
Catalog No. NC9848628
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
35x32 Replacement Thermocouple Probe, Temperature Range: -100 to 500F (-73 to 260C). Shaft Length: 4 in. (102mm).
Catalog No. NC9848628 Supplier Cooper Atkins Corporation Supplier No. 55032
This item has a minimum qty of 2 per supplier requirements.
35x32 Replacement Thermocouple Probe, Temperature Range: -100 to 500F (-73 to 260C). Shaft Length: 4 in. (102mm).