Learn More
Grainger Replacement Electrolyte CLD
Catalog No. 502768019
Encompass_Preferred
The SENSOREX CLDA-4015 is a replacement electrolyte solution for SENSOREX chlorine dioxide analyzers. It is used to refill electrolyte cells in CLDA Series chlorine dioxide analyzers after the initial fill has been depleted. This 500 mL bottle of electrolyte solution is manufactured by SENSOREX to properly replenish electrolyte cells in their chlorine dioxide measurement equipment. The electrolyte solution enables accurate measurement of chlorine dioxide levels in water treatment applications. It has an operating temperature range of 32-122°F.
Catalog No. 50-276-8019 Supplier Grainger Supplier No. 16Z053
The SENSOREX CLDA-4015 is a replacement electrolyte solution for SENSOREX chlorine dioxide analyzers. It is used to refill electrolyte cells in CLDA Series chlorine dioxide analyzers after the initial fill has been depleted. This 500 mL bottle of electrolyte solution is manufactured by SENSOREX to properly replenish electrolyte cells in their chlorine dioxide measurement equipment. The electrolyte solution enables accurate measurement of chlorine dioxide levels in water treatment applications. It has an operating temperature range of 32-122°F.