Learn More
Cole-Parmer Oakton Electrode Electrolyte Powder
Catalog No. 501961778 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-196-1778 Supplier Cole-Parmer Supplier No. 3564070
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.