Learn More
Welch™ Replacement Element for Vacuum Pumps
Catalog No. 01108B
Change view
Click to view available options
Type:
Acid Neutralization Trap
Oil-mist Eliminator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-108B Oil-mist Eliminator
01-104-15A Acid Neutralization Trap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-108B Supplier Welch™ Supplier No. 1417Y05
Add to Cart
Add to Cart
Accessory for Welch vaccum pumps
  • For acid neutralization trap and oil mist elinimator
  • Changes from white to bluish-transparent when spent
Specifications
For Use With Model 1416D, 1400N, 1402N, 1376N hermetically sealed Oil mist eliminator
Type Oil-mist Eliminator