Learn More
Research Products International Corp Replacement Empty Cell Rack with Lid for 248793, 1250 ul Tips, 8/pack
SDP
Catalog No. 501978202
Encompass
Replacement Empty Cell Racks are ideal for storing tips and autoclaving before use. Racks are manufactured from autoclavable polypropylene.
Catalog No. 50-197-8202 Supplier Research Products International Corp Supplier No. 248793RACK
Replacement Empty Cell Racks are ideal for storing tips and autoclaving before use. Racks are manufactured from autoclavable polypropylene.