Learn More
Fisherbrand™ REPLACEMENT FIBERGLASS SLEEVE FOR BEAKER TONGS 4/PK
Catalog No. 13820021 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Fiberglass
Catalog No. Material
13-820-021 Fiberglass
1 options
This item is not returnable. View return policy
Catalog No. 13-820-021
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement fiberglass sleeve covered jaws are designed for use with beaker tongs.
Passivated, and compatible with Gamma/ETO/Autoclavable sterilization methods.
  • Fiberglass construction
  • Passivated
  • Autoclavable
Certifications
ISO 9001-02, GMP, cGMP
Specifications
Includes Sleeves
Product Type Fiberglass Sleeves
For Use With (Application) Replacement sleeves for fiberglass tongs (Fisher part number: 02620)
Material Fiberglass
Quantity 1